Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hvcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042489.2 | NP_001035954.1 | 8 | 2715 | ||
NM_028752.3 | NP_083028.1 | 7 | 2577 | |||
GenBank mRNA | AK002854.1 | - | 7 | 2578 | ||
AK154880.1 | - | 8 | 2607 | |||
AK155123.1 | - | 8 | 2552 | |||
BC021548.1 | - | 8 | 2546 | 4913027 |