Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35f5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028787.4 | NP_083063.2 | 2 | 328 | ||
GenBank mRNA | AK004892.2 | - | 2 | 309 | ||
AK076339.1 | - | 2 | 322 | |||
AK145848.1 | - | 2 | 329 | |||
AK169012.1 | - | 2 | 190 | |||
AK171951.1 | - | 2 | 302 | |||
BC026858.1 | - | 2 | 187 | 4014064 |