Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4932438H23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163695.1 | NP_001157167.1 | 2 | 332 | ||
NM_028905.3 | NP_083181.1 | 3 | 580 | |||
GenBank mRNA | AK016554.1 | - | 3 | 581 | ||
AK035767.1 | - | 2 | 322 | |||
AK043035.1 | - | 3 | 570 | |||
BC032975.1 | - | 1 | 479 | 1264334 |