Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028959.3 | NP_083235.3 | 6 | 782 | ||
GenBank mRNA | AK017134.1 | - | 6 | 774 | ||
AK034892.1 | - | 6 | 775 | |||
AK161033.1 | - | 2 | 107 | |||
AK173189.1 | - | 1 | 691 | |||
BC060686.1 | - | 6 | 745 | |||
BC080681.1 | - | 6 | 738 | |||
BC100549.1 | - | 5 | 632 |