Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scyl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286002.1 | NP_001272931.1 | 14 | 2481 | ||
NM_001286003.1 | NP_001272932.1 | 14 | 2732 | |||
NM_028776.5 | NP_083052.1 | 14 | 2707 | |||
GenBank mRNA | AK004809.2 | - | 14 | 2447 | ||
AK030824.1 | - | 14 | 2682 | |||
AK044100.1 | - | 14 | 2693 | |||
AK047267.1 | - | 2 | 1488 | |||
AK050999.1 | - | 2 | 242 | |||
AK162937.1 | - | 14 | 2482 | |||
BC043085.1 | - | 14 | 2408 | 6417440 | ||
BC066800.1 | - | 14 | 2392 |