Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sipa1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081028.1 | NP_001074497.1 | 17 | 4848 | ||
GenBank mRNA | AK044193.1 | - | 4 | 675 | ||
AK087819.1 | - | 4 | 833 | |||
AK144537.1 | - | 2 | 93 | |||
AK172968.1 | - | 10 | 1591 | |||
BC095969.1 | - | 7 | 1381 | |||
BC116408.1 | - | 2 | 441 | |||
BC128493.1 | - | 2 | 441 | |||
BC144792.1 | - | 4 | 754 | |||
BC145739.1 | - | 4 | 754 | |||
BC152360.1 | - | 2 | 441 | |||
BC152361.1 | - | 2 | 429 |