Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYAA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000394.3 | NP_000385.1 | 1 | 187 | ||
GenBank mRNA | BC113598.1 | - | 1 | 116 | 8322650 | |
BX118596.1 | - | 1 | 187 | |||
CR407691.1 | - | 1 | 95 | |||
KM220588.1 | - | 1 | 145 | |||
KM220590.1 | - | 1 | 152 | |||
KM220591.1 | - | 1 | 111 | |||
KM220592.1 | - | 1 | 153 | |||
U05569.1 | - | 1 | 162 | |||
U66584.1 | - | 1 | 95 | |||
X14789.1 | - | 1 | 108 |
Gene Symbol: LOC102724652 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001314050.2 | NP_001300979.1 | 1 | 154 | ||
GenBank mRNA | BC069528.1 | - | 1 | 145 | 7262133 | |
BF726318.1 | - | 1 | 163 | |||
KM220589.1 | - | 1 | 150 |