Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmbr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029098.3 | NP_083374.1 | 9 | 899 | ||
GenBank mRNA | AK003656.1 | - | 9 | 831 | ||
AK030589.1 | - | 9 | 865 | |||
AK049110.1 | - | 9 | 900 | |||
AK143891.1 | - | 9 | 860 | |||
AK154968.1 | - | 9 | 1033 | |||
AK170328.1 | - | 9 | 860 | |||
AK173121.1 | - | 9 | 1280 | |||
AY052398.1 | - | 9 | 742 | |||
BC023397.1 | - | 9 | 836 | 5043877 |