Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029121.3 | NP_083397.3 | 2 | 333 | ||
GenBank mRNA | AB074757.1 | - | 2 | 320 | ||
AK015654.1 | - | 2 | 320 | |||
BC049547.1 | - | 2 | 321 | 6770428 | ||
BC061059.1 | - | 2 | 333 | 6744043 |