Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171004.1 | NP_001164475.1 | 2 | 1004 | ||
NM_029239.3 | NP_083515.2 | 2 | 1004 | |||
GenBank mRNA | AK138826.1 | - | 2 | 969 | ||
BC027320.1 | - | 1 | 405 | |||
BC037012.1 | - | 1 | 648 | 5342428 | ||
BC089164.1 | - | 2 | 903 | 5400250 |