Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mau2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028993.4 | NP_083269.4 | 13 | 1363 | ||
GenBank mRNA | AK018662.1 | - | 13 | 1331 | ||
AK049277.1 | - | 13 | 1324 | |||
AK081967.1 | - | 13 | 1364 | |||
BC023401.1 | - | 3 | 217 | |||
BC125568.1 | - | 13 | 1363 | 40130083 | ||
BC132139.1 | - | 13 | 1363 | 40130416 |