Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd200r3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128132.1 | NP_001121604.1 | 6 | 1395 | ||
NM_001128133.1 | NP_001121605.1 | 5 | 1176 | |||
GenBank mRNA | AK014671.1 | - | 5 | 1113 | ||
AK046631.1 | - | 2 | 625 |