Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000411.6 | NP_000402.3 | 5 | 554 | ||
NM_001242784.1 | NP_001229713.1 | 5 | 1488 | |||
NM_001242785.1 | NP_001229714.1 | 5 | 647 | |||
XM_005260953.3 | XP_005261010.1 | 4 | 516 | |||
XM_005260955.3 | XP_005261012.1 | 4 | 387 | |||
XM_005260956.3 | XP_005261013.1 | 4 | 563 | |||
XM_006723994.2 | XP_006724057.1 | 4 | 501 | |||
XM_006723995.1 | XP_006724058.1 | 5 | 679 | |||
XM_011529538.1 | XP_011527840.1 | 5 | 534 | |||
XM_011529539.2 | XP_011527841.1 | 5 | 539 | |||
XM_011529540.2 | XP_011527842.1 | 4 | 516 | |||
XM_011529541.2 | XP_011527843.1 | 4 | 390 | |||
XM_017028330.1 | XP_016883819.1 | 4 | 337 | |||
GenBank mRNA | AJ001864.1 | - | 4 | 577 | ||
AK307940.1 | - | 4 | 515 | |||
AK314189.1 | - | 4 | 421 | |||
BC060787.1 | - | 5 | 512 | 30343493 | ||
D23672.2 | - | 5 | 545 | |||
D87328.1 | - | 5 | 1305 | |||
DA334377.1 | - | 5 | 554 | |||
X80160.1 | - | 4 | 405 |