Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIPK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006871.3 | NP_006862.2 | 3 | 404 | ||
GenBank mRNA | AF156884.1 | - | 3 | 194 | ||
AK075275.1 | - | 3 | 358 | |||
AK296140.1 | - | 3 | 357 | |||
AK296309.1 | - | 3 | 339 | |||
AY453693.1 | - | 3 | 194 | |||
AY494982.1 | - | 3 | 194 | |||
AY494983.1 | - | 3 | 194 | |||
BC062584.1 | - | 3 | 335 | 5213027 | ||
DB274842.1 | - | 3 | 404 |