Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029492.4 | NP_083768.4 | 2, 3 | 226 | ||
GenBank mRNA | AK017158.1 | - | 2, 3 | 227 | ||
AK031184.1 | - | 2, 3 | 185 | |||
AK049270.1 | - | 2, 3 | 206 | |||
AK139216.1 | - | 2, 3 | 185 | |||
AK140182.1 | - | 2, 3 | 187 | |||
AK151840.1 | - | 2, 3 | 241 | |||
AK163174.1 | - | 2, 3 | 224 | |||
AK170017.1 | - | 2, 3 | 244 | |||
AK170139.1 | - | 2, 3 | 167 | |||
AY668951.1 | - | 2, 3 | 144 | |||
BC019536.1 | - | 2, 3 | 243 | 4039734 |