Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd14b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029631.3 | NP_083907.3 | 4 | 822 | ||
GenBank mRNA | AK007469.1 | - | 4 | 777 | ||
BC019410.1 | - | 4 | 801 | 4191819 |