Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcgf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029508.3 | NP_083784.1 | 3 | 434 | ||
GenBank mRNA | AK002387.1 | - | 3 | 457 | ||
AK017984.1 | - | 3 | 435 | |||
AK146133.1 | - | 3 | 504 | |||
BC139258.1 | - | 3 | 395 | 8862280 | ||
BC139259.1 | - | 3 | 395 | 8862281 | ||
BC145590.1 | - | 3 | 395 | 9054156 | ||
BC145591.1 | - | 3 | 395 | 9054157 |