Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029582.2 | NP_083858.1 | 9 | 1965 | ||
NM_134105.2 | NP_598866.1 | 2 | 208 | |||
GenBank mRNA | AK139620.1 | - | 9 | 2048 | ||
AK155844.1 | - | 9 | 2019 | |||
AK156154.1 | - | 9 | 2006 | |||
AK157288.1 | - | 9 | 1939 | |||
AK210341.1 | - | 2 | 75 | |||
AK214345.1 | - | 2 | 75 | |||
AK216234.1 | - | 2 | 283 | |||
BC019564.1 | - | 2 | 233 | |||
BC020363.1 | - | 8 | 1412 | |||
BC029643.1 | - | 9 | 1965 | 4010810 |