Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029570.3 | NP_083846.2 | 30 | 3852 | ||
GenBank mRNA | AK003820.1 | - | 3 | 753 | ||
AK129248.1 | - | 30 | 3844 | |||
BC026741.1 | - | 4 | 573 | |||
BC026986.1 | - | 15 | 1921 | |||
BC076603.1 | - | 30 | 3722 | 30629594 | ||
CF952078.1 | - | 3 | 466 |