Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029600.3 | NP_083876.3 | 31 | 4744 | ||
GenBank mRNA | AK006128.1 | - | 4 | 728 | ||
AK149551.1 | - | 31 | 4744 | |||
AK155664.1 | - | 31 | 4722 | |||
AK156615.1 | - | 31 | 4974 | |||
AK172420.1 | - | 31 | 4974 | |||
AY841885.1 | - | 31 | 4725 | |||
BC046560.1 | - | 28 | 4251 | |||
BC048825.1 | - | 31 | 4676 | |||
BC058185.1 | - | 8 | 1193 | |||
BC150788.1 | - | 31 | 4738 | 9087699 |