Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164369.1 | NP_001157841.1 | 3, 4 | 260 | ||
NM_029815.2 | NP_084091.2 | 3, 4 | 260 | |||
GenBank mRNA | AK020254.1 | - | 3, 4 | 262 | ||
AY219233.1 | - | 3, 4 | 243 | |||
BC113166.1 | - | 3, 4 | 240 | 40059981 | ||
BC113957.1 | - | 3, 4 | 240 | 40059979 | ||
BY720653.1 | - | 3, 4 | 262 |