Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168403.1 | NP_001161875.1 | 3 | 687 | ||
NM_001168404.1 | NP_001161876.1 | 3 | 224 | |||
NM_001168405.1 | NP_001161877.1 | 2 | 581 | |||
NM_029898.3 | NP_084174.2 | 2 | 581 | |||
GenBank mRNA | AK039553.1 | - | 2 | 582 | ||
AK045480.1 | - | 2 | 215 | |||
AK081197.1 | - | 3 | 225 | |||
AK138273.1 | - | 2 | 236 | |||
AK140417.1 | - | 3 | 550 | |||
AK155092.1 | - | 1 | 22 | |||
AK171119.1 | - | 1 | 22 | |||
BC152317.1 | - | 2 | 208 | 40088531 | ||
BC152536.1 | - | 2 | 208 | 40088530 |