Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029933.4 | NP_084209.3 | 10 | 4081 | ||
GenBank mRNA | AK020724.1 | - | 1 | 75 | ||
AK084676.1 | - | 3 | 2008 | |||
AK147659.1 | - | 10 | 4081 | |||
AY296061.1 | - | 7 | 3597 | |||
BC019641.1 | - | 2 | 528 | |||
BC137850.1 | - | 7 | 3569 | 8860869 |