Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mboat7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029934.3 | NP_084210.2 | 5 | 1180 | ||
GenBank mRNA | AB055410.1 | - | 5 | 1178 | ||
AK019981.1 | - | 5 | 591 | |||
AK038998.1 | - | 5 | 602 | |||
AK044149.1 | - | 5 | 607 | |||
AK150010.1 | - | 5 | 574 | |||
AK154606.1 | - | 5 | 1179 | |||
AK158998.1 | - | 4 | 935 | |||
AK170124.1 | - | 5 | 657 | |||
BC023417.1 | - | 5 | 628 | 5028759 | ||
BC118958.2 | - | 5 | 825 | 40093821 | ||
EU016380.1 | - | 4 | 392 |