Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001326.1 | NP_001001326.1 | 6 | 1829 | ||
NM_029811.2 | NP_084087.2 | 5 | 578 | |||
GenBank mRNA | AK158270.1 | - | 6 | 1834 | ||
BC030391.1 | - | 2 | 304 | 4950637 | ||
BC051034.1 | - | 6 | 2010 | 5345299 | ||
CF737707.1 | - | 3 | 311 |