Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs7bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029879.2 | NP_084155.2 | 4 | 1306 | ||
GenBank mRNA | AK028323.1 | - | 4 | 683 | ||
AK032138.1 | - | 4 | 1306 | |||
AK036240.1 | - | 4 | 1213 | |||
AK043529.1 | - | 1 | 110 | |||
AK046733.1 | - | 4 | 935 | |||
AK137417.1 | - | 4 | 1180 | |||
BC118957.2 | - | 4 | 838 | 40093423 | ||
DQ104214.1 | - | 4 | 596 |