Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029963.2 | NP_084239.1 | 4 | 353 | ||
GenBank mRNA | AB049941.1 | - | 4 | 353 | ||
AK018829.1 | - | 4 | 361 | |||
AK078198.1 | - | 4 | 361 | |||
AK083366.1 | - | 4 | 355 | |||
AK155101.1 | - | 4 | 369 | |||
BC021947.1 | - | 4 | 351 | 5098933 |