Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ermn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029972.3 | NP_084248.2 | 1 | 164 | ||
GenBank mRNA | AK020733.1 | - | 1 | 276 | ||
AK078089.1 | - | 1 | 254 | |||
AK136820.1 | - | 1 | 254 | |||
AK137305.1 | - | 1 | 256 | |||
AK220372.1 | - | 1 | 283 | |||
BC089515.1 | - | 1 | 165 | 6589784 | ||
BC090670.1 | - | 1 | 196 | 30941185 | ||
DQ334270.1 | - | 1 | 142 |