Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csgalnact2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030165.3 | NP_084441.3 | 2 | 576 | ||
GenBank mRNA | AK019496.1 | - | 2 | 579 | ||
AK028045.1 | - | 2 | 557 | |||
AK145992.1 | - | 2 | 563 | |||
BC060169.1 | - | 1 | 472 |