Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wibg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170869.2 | NP_001164340.1 | 2, 3 | 206 | ||
NM_001253704.1 | NP_001240633.1 | 3, 4 | 261 | |||
NM_030100.4 | NP_084376.2 | 2, 3 | 214 | |||
GenBank mRNA | AJ459407.1 | - | 2, 3 | 131 | ||
AK020702.1 | - | 2, 3 | 205 | |||
BC049647.1 | - | 2, 3 | 139 | 6707378 | ||
BI989958.1 | - | 2, 3 | 206 | |||
BQ177652.1 | - | 2, 3 | 133 | |||
CK332572.1 | - | 2, 3 | 155 |