Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpe65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029987.2 | NP_084263.2 | 4 | 350 | ||
GenBank mRNA | AF410461.1 | - | 3 | 298 | ||
AK020906.1 | - | 4 | 349 | |||
BC090656.1 | - | 4 | 326 | |||
BC130028.1 | - | 3 | 302 | 40140883 | ||
CJ133618.1 | - | 4 | 351 |