Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nadsyn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ305343.1 | - | 20 | 2012 | ||
AK020264.1 | - | 20 | 2012 | |||
AK041101.1 | - | 20 | 1999 | |||
AK042389.1 | - | 20 | 1972 | |||
BC018248.1 | - | 18 | 1778 | |||
BC038016.1 | - | 20 | 1958 | 3589586 |