Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030566.2 | NP_085043.2 | 3 | 673 | ||
GenBank mRNA | AK029188.1 | - | 1 | 33 | ||
AK089938.1 | - | 3 | 377 | |||
AK144836.1 | - | 3 | 359 | |||
AK157832.1 | - | 3 | 334 | |||
BC004088.1 | - | 2 | 419 | 3593722 | ||
BC015287.1 | - | 3 | 681 | 4240442 | ||
BU514356.1 | - | 3 | 673 |