Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klrb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030599.4 | NP_085102.4 | 6 | 677 | ||
GenBank mRNA | AF324826.1 | - | 6 | 653 | ||
AF338321.1 | - | 6 | 608 | |||
AF338322.1 | - | 6 | 608 | |||
AF342896.1 | - | 6 | 656 | |||
AK030702.1 | - | 6 | 626 | |||
AK138766.1 | - | 5 | 560 | |||
BC055379.1 | - | 6 | 676 | 30076215 | ||
BC100580.1 | - | 6 | 664 | 30074482 |