Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KALRN SIRNA Assays ›
Gene Symbol:
KALRN
Gene Name:
kalirin, RhoGEF kinase
Gene Aliases:
ARHGEF24, CHD5, CHDS5, DUET, DUO, HAPIP, TRAD
Chromosome Location:
Chr.3: 124033341 - 124726325 on Build GRCh38
UniGene ID:
Hs.13281
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
847
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KALRN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001024660.4
NP_001019831.2
35
5511
NM_001322988.1
NP_001309917.1
35
5511
NM_001322993.1
NP_001309922.1
2
426
NM_001322994.1
NP_001309923.1
2
426
NM_001322995.1
NP_001309924.1
2
426
NM_001322996.1
NP_001309925.1
2
426
NM_001322997.1
NP_001309926.1
2
426
NM_001322998.1
NP_001309927.1
2
426
NM_001322999.1
NP_001309928.1
2
426
NM_001323000.1
NP_001309929.1
2
426
NM_001323001.1
NP_001309930.1
2
426
NM_007064.4
NP_008995.2
2
426
XM_006713810.3
XP_006713873.1
35
5756
XM_006713811.3
XP_006713874.1
35
5729
XM_006713812.3
XP_006713875.1
35
5717
XM_006713813.3
XP_006713876.1
35
5690
XM_006713814.3
XP_006713877.1
35
5756
XM_011513279.2
XP_011511581.1
35
5756
XM_011513280.2
XP_011511582.1
35
5757
XM_011513281.2
XP_011511583.1
35
28378
XM_011513283.2
XP_011511585.1
35
5753
XM_011513285.2
XP_011511587.1
35
5752
XM_017007429.1
XP_016862918.1
35
5756
XM_017007430.1
XP_016862919.1
35
5756
XM_017007431.1
XP_016862920.1
35
5752
XM_017007432.1
XP_016862921.1
35
5752
GenBank mRNA
AB011422.1
-
2
380
AK125979.1
-
3
637
BC058015.1
-
2
406
DB450617.1
-
2
426
Back To Top
More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
signal transduction
nervous system development
vesicle-mediated transport
regulation of Rho protein signal transduction
intracellular signal transduction
positive regulation of apoptotic process
positive regulation of GTPase activity
ephrin receptor signaling pathway
regulation of small GTPase mediated signal transduction
protein serine/threonine kinase activity
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
GTPase activator activity
ATP binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0