Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim34a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030684.3 | NP_109609.2 | 2 | 590 | ||
GenBank mRNA | AF220139.1 | - | 2 | 487 | ||
AF220140.1 | - | 490 | ||||
AK089187.1 | - | 605 | ||||
AK171840.1 | - | 2 | 550 | |||
AK172135.1 | - | 2 | 550 |
Gene Symbol: Trim34b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243916.1 | NP_001230845.1 | 1 | 331 | ||
GenBank mRNA | BC145515.1 | - | 1 | 331 |