Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sacm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030692.4 | NP_109617.1 | 5, 6 | 498 | ||
GenBank mRNA | AJ245720.1 | - | 5, 6 | 628 | ||
AJ297526.1 | - | 5, 6 | 499 | |||
AK089174.1 | - | 5, 6 | 582 | |||
AK122390.1 | - | 5, 6 | 563 | |||
AK154759.1 | - | 5, 6 | 598 | |||
AK161111.1 | - | 5, 6 | 532 | |||
AK171080.1 | - | 5, 6 | 582 | |||
BC117752.1 | - | 5, 6 | 499 | 40090556 | ||
BC117753.1 | - | 5, 6 | 499 | 40090558 |