Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000429.2 | NP_000420.1 | 3, 4 | 554 | ||
XM_005269842.4 | XP_005269899.1 | 4, 5 | 784 | |||
GenBank mRNA | AK290820.1 | - | 3, 4 | 420 | ||
BC018359.1 | - | 3, 4 | 521 | 4341915 | ||
BM738684.1 | - | 3, 4 | 566 | |||
D49357.1 | - | 3, 4 | 317 | |||
X69078.1 | - | 3, 4 | 370 |