Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000432.3 | NP_000423.2 | 3 | 198 | ||
GenBank mRNA | AF020768.1 | - | 2 | 132 | ||
AK312013.1 | - | 3 | 175 | |||
BC015821.1 | - | 3 | 180 | 4245364 | ||
BC031006.1 | - | 3 | 175 | 4292208 | ||
BC031008.1 | - | 3 | 177 | 4297454 | ||
CR456962.1 | - | 2 | 127 | |||
CR541957.1 | - | 2 | 127 | |||
M22815.1 | - | 3 | 149 | |||
S69022.1 | - | 3 | 204 | |||
X14332.1 | - | 2 | 127 | |||
X57542.1 | - | 3 | 149 | |||
X66141.1 | - | 3 | 157 |