Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114386.1 | NP_001107858.1 | 8 | 772 | ||
NM_031881.2 | NP_114087.2 | 7 | 821 | |||
GenBank mRNA | AF277232.1 | - | 7 | 454 | ||
AK004969.1 | - | 7 | 514 | |||
AK031842.1 | - | 7 | 502 | |||
AK042621.1 | - | 8 | 455 | |||
AK122283.1 | - | 7 | 624 | |||
AK163497.1 | - | 6 | 302 | |||
AK183418.1 | - | 2 | 154 | |||
AK188093.1 | - | 3 | 154 | |||
AK191211.1 | - | 3 | 154 | |||
AK198307.1 | - | 3 | 154 | |||
AK199964.1 | - | 3 | 154 | |||
AK202185.1 | - | 3 | 154 | |||
AK205175.1 | - | 3 | 154 | |||
BC039746.1 | - | 7 | 465 | 3157137 | ||
BC071210.1 | - | 7 | 442 | 4972426 | ||
BC082281.1 | - | 7 | 491 |