Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000342.3 | NP_000333.1 | 20 | 3359 | ||
XM_005257593.4 | XP_005257650.1 | 18 | 3127 | |||
XM_011525129.1 | XP_011523431.1 | 19 | 3296 | |||
GenBank mRNA | M27819.1 | - | 20 | 3324 | ||
X12609.1 | - | 20 | 3360 |