Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000355.3 | NP_000346.2 | 7, 8 | 1356 | ||
NM_001184726.1 | NP_001171655.1 | 7, 8 | 1275 | |||
GenBank mRNA | AK225445.1 | - | 7, 8 | 1145 | ||
AK313711.1 | - | 7, 8 | 1246 | |||
BC001176.1 | - | 7, 8 | 1234 | 3353271 | ||
BC011239.1 | - | 7, 8 | 1154 | 4153362 | ||
CR456591.1 | - | 7, 8 | 1146 | |||
L02647.1 | - | 7, 8 | 1204 | |||
L02648.1 | - | 7, 8 | 1160 | |||
M60396.1 | - | 7, 8 | 1144 |