Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033444.2 | NP_254279.1 | 5 | 627 | ||
GenBank mRNA | AK088809.1 | - | 5 | 438 | ||
AK150382.1 | - | 5 | 905 | |||
AK150611.1 | - | 5 | 616 | |||
AK153509.1 | - | 5 | 905 | |||
AK167765.1 | - | 5 | 633 | |||
AK168262.1 | - | 5 | 447 | |||
AK186702.1 | - | 3 | 201 | |||
BC004658.1 | - | 5 | 627 | 3498118 |