Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubqln4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033526.2 | NP_277068.1 | 1 | 166 | ||
GenBank mRNA | AB040050.1 | - | 1 | 166 | ||
AK150211.1 | - | 1 | 104 | |||
AK156515.1 | - | 1 | 119 | |||
AK160578.1 | - | 1 | 118 | |||
BC017686.1 | - | 1 | 128 | 4236887 |