Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033541.4 | NP_291019.1 | 3 | 586 | ||
GenBank mRNA | AB067528.1 | - | 2 | 243 | ||
AF459815.1 | - | 2 | 243 | |||
AF459816.1 | - | 2 | 243 | |||
AK028986.1 | - | 3 | 587 | |||
AK078952.1 | - | 2 | 254 | |||
AK163253.1 | - | 2 | 259 | |||
BC108955.1 | - | 2 | 252 | 40046017 | ||
BC126877.1 | - | 2 | 251 | 40046021 |