Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aff4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033565.2 | NP_291043.1 | 3 | 564 | ||
GenBank mRNA | AF190449.1 | - | 3 | 506 | ||
AK044058.1 | - | 3 | 448 | |||
AK054401.1 | - | 3 | 412 | |||
BC099959.1 | - | 2 | 231 | |||
BC138999.1 | - | 3 | 237 | 8862021 | ||
BC145490.1 | - | 3 | 237 | 9054053 |