Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000370.3 | NP_000361.1 | 5 | 1027 | ||
XM_006716468.3 | XP_006716531.1 | 3 | 1149 | |||
GenBank mRNA | BC041784.1 | - | 5 | 1013 | ||
BC058000.1 | - | 5 | 1031 | 4610804 | ||
D49488.1 | - | 5 | 1013 |