Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TESK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007170.2 | NP_009101.2 | 2 | 484 | ||
XM_005270355.3 | XP_005270412.1 | 2 | 456 | |||
XM_006710287.2 | XP_006710350.1 | 3 | 524 | |||
XM_011540497.1 | XP_011538799.1 | 4 | 557 | |||
XM_011540500.2 | XP_011538802.1 | 2 | 456 | |||
XM_017000047.1 | XP_016855536.1 | 3 | 689 | |||
XM_017000048.1 | XP_016855537.1 | 2 | 658 | |||
XM_017000049.1 | XP_016855538.1 | 2 | 658 | |||
GenBank mRNA | AB057597.1 | - | 2 | 476 | ||
AJ132545.1 | - | 2 | 476 | |||
AK027573.1 | - | 2 | 458 | |||
AK299269.1 | - | 2 | 479 | |||
AK302494.1 | - | 2 | 516 | |||
BC022194.1 | - | 2 | 464 | |||
BC033085.1 | - | 1 | 95 | 5425494 | ||
DA990759.1 | - | 3 | 524 | |||
DB161528.1 | - | 4 | 557 |