Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033444.2 | NP_254279.1 | 6 | 1083 | ||
GenBank mRNA | AK088809.1 | - | 6 | 894 | ||
AK150382.1 | - | 6 | 1361 | |||
AK150611.1 | - | 6 | 1072 | |||
AK153509.1 | - | 6 | 1361 | |||
AK167765.1 | - | 6 | 1089 | |||
AK168262.1 | - | 6 | 903 | |||
AK205498.1 | - | 1 | 41 | |||
BC004658.1 | - | 6 | 1083 | 3498118 |
Gene Symbol: Sept9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC094455.1 | - | 3116 |