Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Laptm4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033521.3 | NP_277056.1 | 7 | 1660 | ||
GenBank mRNA | AK077269.1 | - | 7 | 1523 | ||
AK077933.1 | - | 7 | 1660 | |||
AK079581.1 | - | 7 | 1523 | |||
AK082807.1 | - | 7 | 1539 | |||
AK146868.1 | - | 6 | 1427 | |||
AK166014.1 | - | 7 | 1562 | |||
AK207094.1 | - | 1 | 149 | |||
AK209001.1 | - | 1 | 149 | |||
BC019120.1 | - | 7 | 1530 | 4507607 | ||
EU561010.1 | - | 7 | 1660 |